Chlorhexidin ≥85%, weißgraues Puder

Lieferant: MP Biomedicals
Gefahr

Synonyme: 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide]

0219135805
ICNA0219135805EA 404 EUR
ICNA0219135805
Chlorhexidin ≥85%, weißgraues Puder
Chlorhexidin

Chlorhexidine is a chemical antiseptic. It is effective on both Gram-positive and Gram-negative bacteria, although it is less effective with some Gram-negative bacteria. It has both bactericidal and bacteriostatic mechanisms of action.


An intermediate for the production of chlorhexidine salts which are used as an antiseptic and disinfectant agent. It is effective against a wide range of bacteria, some fungi and some viruses, and an agent for the prevention of gingivitis. Commercial ophthalmic products have used this agent to replace thimerosal as a preservative; however, it can cause skin irritation. It ia used in analytical standard for pharmaceuticals.


An intermediate for the production of chlorhexidine salts which are used as an antiseptic and disinfectant agent. It is effective against a wide range of bacteria, some fungi and some viruses, and an agent for the prevention of gingivitis. Commercial ophthalmic products have used this agent to replace thimerosal as a preservative; however, it can cause skin irritation. It ia used in analytical standard for pharmaceuticals.


Slightly soluble in water (0.08% w/v).


Bitte beachten Research Use Only (RUO).

Formel: C₂₂H₃₀Cl₂N₁₀
Molecular Weight: 505,4466 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00009673
CAS-Nummer: 55-56-1
EINECS: 200-238-7

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Assay ≥85%
FTIR Conforms to Reference
Residue on Ignition ≤0,2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR