Ammoniumacetat, wasserfrei, Pulver ACS
Lieferant: MP Biomedicals
Synonyme:
Essigsäure Ammoniumsalz
0219140480
0219140490
0219140491
ICNA0219140480EA
33
EUR
ICNA0219140480
ICNA0219140490
ICNA0219140491
Ammoniumacetat, wasserfrei, Pulver ACS
Ammoniumacetat
Formel:
H₃CCOONH₄ |
MDL:
MFCD00013066 CAS-Nummer: 631-61-8 |
|
Spezifikation Testergebnisse
Synonyms | Acetic acid ammonium salt |
Form | White Crystalline Powder |
Storage | Store at Room Temperature (15 - 30 °C), Desiccate |
Assay | ≥97% |
Heavy Metals | ≤5 ppm |
Insoluble Matter | ≤0,005% |
pH | 6,7 - 7,3 (5% aq soln) |
Residue on Ignition | ≤0,01% |
Chloride (Cl) | ≤5 ppm |
Nitrate (NO3) |
|
Sulfate (SO4) | ≤0,001% |
Iron (Fe) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...