Ammoniumacetat, wasserfrei, Pulver ACS

Lieferant: MP Biomedicals

Synonyme: Essigsäure Ammoniumsalz

0219140480 0219140490 0219140491
ICNA0219140480EA 33 EUR
ICNA0219140480 ICNA0219140490 ICNA0219140491
Ammoniumacetat, wasserfrei, Pulver ACS
Ammoniumacetat
Formel: H₃CCOONH₄
MDL: MFCD00013066
CAS-Nummer: 631-61-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Synonyms Acetic acid ammonium salt
Form White Crystalline Powder
Storage Store at Room Temperature (15 - 30 °C), Desiccate
Assay ≥97%
Heavy Metals ≤5 ppm
Insoluble Matter ≤0,005%
pH 6,7 - 7,3 (5% aq soln)
Residue on Ignition ≤0,01%
Chloride (Cl) ≤5 ppm
Nitrate (NO3) 0,001%
Sulfate (SO4) ≤0,001%
Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR