Ammoniumsulfat ≥99%, Pulver ACS

Lieferant: MP Biomedicals

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

02191409.5 0219140901 0219140905
ICNA02191409.5EA 61.7 EUR
ICNA02191409.5 ICNA0219140901 ICNA0219140905
Ammoniumsulfat ≥99%, Pulver ACS
Ammoniumsulfat

Ammonium Sulfate may be used for the precipitation or fractionation of proteins or for purification of antibodies. Useful for crystallographic analysis of nucleic acids and proteins. The use of ammonium sulfate in long PCR buffer, in PCR lysis solution, and in second strand cDNA synthesis has been reported.


Soluble freely in water; insoluble in alcohol, acetone.


Bitte beachten Research Use Only (RUO).

Formel: (NH₄)₂SO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2

Bestellen

Spezifikation Testergebnisse

Assay ≥99%
pH (5% aq soln) 5 - 6
Insoluble Matter ≤0.005%
Residue on Ignition ≤0,005%
Chloride (Cl) ≤5 ppm
Nitrate (NO₃) ≤0,001%
Phosphate (PO4) ≤5 ppm
Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR