Kaliumdihydrogenphosphat, wasserfrei ≥99%, weißes kristallines Pulver ACS

Lieferant: MP Biomedicals

02191430.1 02191430.5 0219143001 0219143005
ICNA02191430.1EA 35.2 EUR
ICNA02191430.1 ICNA02191430.5 ICNA0219143001 ICNA0219143005
Kaliumdihydrogenphosphat, wasserfrei ≥99%, weißes kristallines Pulver ACS
Kaliumdihydrogenphosphat

Potassium phosphate is a reagent with high buffering capacity. It occurs in several forms: monobasic, dibasic, and tribasic (K3PO4). Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.


Potassium Phosphate Monobasic is used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.


Soluble in water (0,5 g in 10 ml water), insoluble to slightly soluble in ethanol; insoluble in alcohol.


Bitte beachten Research Use Only (RUO).

Formel: KH₂PO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Assay ≥99%
pH (5% aq soln, 25 °C ) 4,1 - 4,5
Insoluble Matter ≤0,01%
Loss on Drying (105 °C) ≤0,2%
Chloride (Cl) ≤0,001%
Sulphate (SO₄) ≤0,003%
Heavy Metals (as Pb) <0,001%
Iron (Fe) ≤0,002%
Sodium (Na) ≤0,005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR