Natriumhydrogencarbonat 99,7-100,3% (Trockenmasse), weißes Pulver ACS

Lieferant: MP Biomedicals

02191435.5 0219143501
ICNA02191435.5EA 39.9 EUR
ICNA02191435.5 ICNA0219143501
Natriumhydrogencarbonat 99,7-100,3% (Trockenmasse), weißes Pulver ACS
Natriumhydrogencarbonat

Soluble in Water.

Formel: NaHCO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00003528
CAS-Nummer: 144-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Assay 99,7 - 100,3% (dried basis)
Insoluble Matter ≤0,015%
Chloride (Cl) <0,003%
Phosphate (PO4) <0,001%
Sulphur Compounds (as SO4) <0,003%
Ammonium (NH4) <5 ppm
Magnesium (Mg) <0,005%
Potassium (K) <0,005%
Calcium (Ca) <0,02%
Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) <0,001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR