di-Natriumoxalat ≥99,5%, Pulver ACS

Lieferant: MP Biomedicals
Achtung

0219143991 0219143980 0219143990
ICNA0219143991EA 251 EUR
ICNA0219143991 ICNA0219143980 ICNA0219143990
di-Natriumoxalat ≥99,5%, Pulver ACS
di-Natriumoxalat

Soluble in water.


Chloride ≤0,002% ; sulphate ≤0,002% ; Ammonium ≤0,002%; Potassium ≤0,005%

Formel: Na₂C₂O₄
Schmelzpunkt: 250…270 °C
Dichte: 2,27 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012465
CAS-Nummer: 62-76-0
EINECS: 200-550-3
Merck Index: 13,08723

Bestellen

Spezifikation Testergebnisse

Assay ≥99.5%
Insoluble Matter ≤0.005%
Loss on Drying ≤0.01%
Neutrality Passes
Chloride ; ≤0.002%
Sulfate ≤0.002%
Ammonium ≤0.002%
Heavy Metals ≤0.002%
Iron ≤0.001%
Potassium ≤0.005%
Substances Darkened by Hot Sulfuric Acid Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR