Anthralin 99, powder_yellow
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-on, 1,8,9-Anthracentriol, 1,8,9-Trihydroxyanthracen
Dithranol (1, 8 - dihydroxy-9-anthrone) is also known as anthralin. It is a vital topical antipsoriatic drugs. Dithranol, an athracene derivative, is a free radical generating mitochondrial poison that induces apoptosis in keratinocytes.
Dithranol, a potential anti-psoriasis medication, may be used to study its safety, efficacy, metabolism and pharmacological profile. Dithranol may be used to study is physicochemical properties, as a reference compound and to develop effective drug delivery vehicles. Dithranol may be used as a matrix for matrix assisted laser desorption/ionization imaging on a Fourier transform ion cyclotron resonance mass spectrometer.
Dithranol, an athracene derivative, is a free radical generating mitochondrial poison that induces apoptosis in keratinocytes. Dithranol is being studied as a potential anti-psoriasis medication.
Soluble in chlororform (20 mg/ml - clear, bright yellow solution) and in fixed oilds; slightly soluble in ethanol (95%) and in ether; practically insoluble in water.
Formel:
C₁₄H₁₀O₃ Molecular Weight: 226.23 g/mol Siedepunkt: 400 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053409 CAS-Nummer: 1143-38-0 EINECS: 214-538-0 Merck Index: 12,00723 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
Melting Point | 175 - 182°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...