Anthralin 99, powder_yellow

Lieferant: MP Biomedicals
Achtung

Synonyme: Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-on, 1,8,9-Anthracentriol, 1,8,9-Trihydroxyanthracen

0219150294 0219150283 0219150291
ICNA0219150294EA 473 EUR
ICNA0219150294 ICNA0219150283 ICNA0219150291
Anthralin 99, powder_yellow
Anthralin

Dithranol (1, 8 - dihydroxy-9-anthrone) is also known as anthralin. It is a vital topical antipsoriatic drugs. Dithranol, an athracene derivative, is a free radical generating mitochondrial poison that induces apoptosis in keratinocytes.


Dithranol, a potential anti-psoriasis medication, may be used to study its safety, efficacy, metabolism and pharmacological profile. Dithranol may be used to study is physicochemical properties, as a reference compound and to develop effective drug delivery vehicles. Dithranol may be used as a matrix for matrix assisted laser desorption/ionization imaging on a Fourier transform ion cyclotron resonance mass spectrometer.


Dithranol, an athracene derivative, is a free radical generating mitochondrial poison that induces apoptosis in keratinocytes. Dithranol is being studied as a potential anti-psoriasis medication.


Soluble in chlororform (20 mg/ml - clear, bright yellow solution) and in fixed oilds; slightly soluble in ethanol (95%) and in ether; practically insoluble in water.

Formel: C₁₄H₁₀O₃
Molecular Weight: 226.23 g/mol
Siedepunkt: 400
Lagertemperatur: Raumtemperatur
MDL: MFCD00053409
CAS-Nummer: 1143-38-0
EINECS: 214-538-0
Merck Index: 12,00723

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Melting Point 175 - 182°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR