D(-)-N-Methylglucamin, powder_white

Lieferant: MP Biomedicals

Synonyme: (-)-N-Methyl-D-glucamin, (-)-1-Methylamino-1-desoxy-D-glucitol, Meglumin, (-)-1-Desoxy-1-(methylamino)-D-glucitol

0219150690 0219150691
ICNA0219150690EA 125 EUR
ICNA0219150690 ICNA0219150691
D(-)-N-Methylglucamin, powder_white
D(-)-N-Methylglucamin

N-Methylglucamine is used in the synthesis of surface active agents, pharmaceuticals, dyes; an antimonate as antiprotozoal. Used as a Na+ substitute in systems to maintain physiological solutions as isotonic but without sodium. Has also been used in the studies of rabbit cerebral artery, rabbit cardiac myocytes, perfused rat intestine, and frog skeletal muscle.


Soluble in water (50 mg/ml - clear, colourless solution).

Formel: H₃CNHCH₂(CH(OH))₄CH₂OH
Molecular Weight: 195.22 g/mol
Schmelzpunkt: 125…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004707
CAS-Nummer: 6284-40-8
Merck Index: 12,06154

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR