Kaliumiodid, powder_crystalline_white

Lieferant: MP Biomedicals
Achtung

Synonyme: Iodkalium

0219151380 0219151390 0219151391
ICNA0219151380EA 0 EUR
ICNA0219151380 ICNA0219151390 ICNA0219151391
Kaliumiodid, powder_crystalline_white
Kaliumiodid

Potassium Iodide is known as Potassium Salt of Hydriodic Acid [HI], available in white crystalline powder form. It is a mild reducing agent and can be easily oxidized to Iodine by powerful oxidizing agent like chlorine. With elemental iodine it forms I3 (tri iodide), the salts of tri iodide are highly water soluble.
Readily soluble in water.


Potassium iodide is used in organic synthesis for the preparation of Aryl iodides and Iodo alkanes. It is used in biochemistry, often as a fluorescence quenching agent which acts by collisional quenching of fluorescent substances by the iodide ion. It is also used in extraction of single stranded DNA.

Formel: Kl
Molecular Weight: 166 g/mol
Siedepunkt: 1330
Schmelzpunkt: 681 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011405
CAS-Nummer: 7681-11-0
Merck Index: 13,07727

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
pH (5% aq soln) 6 - 9,2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR