Chloroquin-diphosphat (Salz)

Lieferant: MP Biomedicals

Synonyme: N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate

0219391910 0219391925 0219391950 0219391980 0219391983
ICNA0219391910EA 33.9 EUR
ICNA0219391910 ICNA0219391925 ICNA0219391950 ICNA0219391980 ICNA0219391983
Chloroquin-diphosphat (Salz)
Chloroquine diphosphate salt

Chloroquine Diphosphate salt is member of quinolone family, Chloroquine is a lysosomotropic agent (accumulate inside the acidic parts of the cell), this accumulation leads to inhibition of lysosomal enzymes that require an acidic pH, and prevents fusion of endosomes and lysosomes. Standard anti-malarial drug. Substrate for MRP in multidrug resistant cell line and inhibits photoaffinity labeling of MRP by quinoline-based photoactive drug IAAQ (N-[4-[1-hydroxy-2-(dibutylamino)ethyl]quinolin-8-yl]-4-azidosalicylamide).


  • Solubility:
  • Soluble in water. Practically insoluble in alcohol, benzene, chloroform and ether.
  • Typical Working Concentration:
  • Chloroquine showed very high antiviral activity against NiV but very little activity against the other viruses at concentrations lower than 20 µM


Chloroquine diphosphate salt is used to study the role of endosomal acidification in cellular processes, such as the signaling of intracellular TLRs. It can be used as DNA intercalator & to dissociate antigen antibody complexes without denaturing red blood cell antigens. DNA intercalator. Also used to increase transfection efficiency.

Formel: C₁₈H₂₆ClN₃
Molecular Weight: 515.86 g/mol
Schmelzpunkt: 200 °C (Zers., Lit.)
MDL: MFCD00069852
CAS-Nummer: 50-63-5
EINECS: 200-055-2

Bestellen

Spezifikation Testergebnisse

Physical Appearance Off-White Powder
pH (1% aq soln) 4.0 - 5.0
pKa 8.4 and 10.8 (for chloroquine base at 20 °C)
Solubility Soluble in water. Practically insoluble in alcohol, benzene, chloroform and ether.
Typical Working Concentration Chloroquine showed very high antiviral activity against NiV but very little activity against the other viruses at concentrations lower than 20 µM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR