D(+)-Glucose, weißes Pulver, Molekularbiologische Qualität

Lieferant: MP Biomedicals

0219402483 0219402491
ICNA0219402483EA 27 EUR
ICNA0219402483 ICNA0219402491
D(+)-Glucose, weißes Pulver, Molekularbiologische Qualität
D(+)-Glucose

D-(+)-Glucose is ideal as a culture media component.


Bitte beachten Research use only (RUO).

Formel: C₆H₁₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00148912
CAS-Nummer: 50-99-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Optical Rotation (c=10, Water, NH4OH) +52,5 ± 5°
Moisture Report Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR