Gentamicinsulfat

Lieferant: MP Biomedicals
Gefahr

Synonyme: Gentamicin Sulfatsalz

02194530.6 0219453005 0219453001
ICNA02194530.6EA 41.8 EUR
ICNA02194530.6 ICNA0219453005 ICNA0219453001
Gentamicinsulfat
Gentamicinsulfat

Gentamicin is an aminoglycoside antibiotic complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is a mixture of 3 major components designated as C1, C1a, and C2. Gentamicin is used as the sulfate salt. Each component consists of five basic nitrogens and requires five equivalents of sulfuric acid per mole of gentamicin base. Mode of action: Gentamicin causes codon misreading by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site.The bactericidal effect of gentamicin on Pseudomonas aeruginosa is exerted by the binding of gentamicin to the outer membrane, where it displaces natural cations, destabilizes the membrane, and forms holes in the cell surface.Antimicrobial spectrum: Gram-negative bacteria, Staphylococcus aureus and other Gram-positive bacteria. Gentamicin sulfate is a broad spectrum antibiotic. It inhibits the growth of a wide variety of Gram-positive and Gram-negative microorganisms, including strains resistant to tetracycline, chloramphenicol, kanamycin, and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus, and Streptococcus. Gentamicin sulfate inhibits bacterial protein biosynthesis by binding to the 30S subunit of the ribosome.


  • Gentamicin sulfate is soluble in water (50 mg/mL), yielding a clear to very slightly hazy, colorless to faint. It is practically insoluble in alcohol and other organic solvents.


Storage temperature: +4 °C.

Schmelzpunkt: 218…237 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00270181
CAS-Nummer: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR