D(+)-Glucose, wasserfrei, weißes Pulver Reagenz für die Zellkultur

Lieferant: MP Biomedicals

02194672.1 0219467201 0219467205
ICNA02194672.1EA 46.1 EUR
ICNA02194672.1 ICNA0219467201 ICNA0219467205
D(+)-Glucose, wasserfrei, weißes Pulver Reagenz für die Zellkultur
D(+)-Glucose

Storage: Store at Room Temperature (15-30 °C).
D- (+)-Glucose is a monosaccharide. It is a mixture of α- and β-anomers, primarily the α-anomer.
D- (+)-Glucose, anhydrous is a main source of energy for living organisms. It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen.


  • Anhydrous
  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Soluble in 20% aqueous clear colorless solution.

Formel: C₆H₁₂O₆
Schmelzpunkt: 150…170 °C (Lit.)
Dichte: 1,544 g/mL
Lagertemperatur: Raumtemperatur
MDL: MFCD00148912
CAS-Nummer: 50-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Optical Rotation (c=10, Water, NH4OH) +52.5 ± 5°
Moisture ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR