L(+)-Leucin

Lieferant: MP Biomedicals

Synonyme: H-Leu-OH, (S)-(+)-α-Amino-γ-methylvaleriansäure, L-Leucin, (S)-(+)-Leucin, (S)-(+)-2-Amino-4-methylpentansäure

0219469483
ICNA0219469483EA 279 EUR
ICNA0219469483
L(+)-Leucin
L(+)-Leucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131,17 g/mol
Schmelzpunkt: 300 °C
Dichte: 1,165 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002617
CAS-Nummer: 61-90-5
EINECS: 200-522-0
Merck Index: 14,05451

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR