EDTA Dinatriumsalz Dihydrat ≥99%, weißes kristallines Pulver für die Molekularbiologie

Lieferant: MP Biomedicals

0219482250 0219482280 0219482283 0219482291
ICNA0219482250EA 25.9 EUR
ICNA0219482250 ICNA0219482280 ICNA0219482283 ICNA0219482291
EDTA Dinatriumsalz Dihydrat ≥99%, weißes kristallines Pulver für die Molekularbiologie
EDTA Dinatriumsalz Dihydrat

Storage: Store at Room Temperature (15-30 °C).
Ethylenediamine Tetraacetic Acid is a polyamino carboxylic acid hexadentate ligand and a chelating agent. This product is designated as Molecular Biology grade and is suitable for molecular biology applications.
Ethylenediamine Tetraacetic Acid is a chelator of divalent cations. It also inhibits enzymes, such as metalloproteases, that require divalent cations for activity.


  • Molecular Biology Reagent Grade
  • Presentation: White Crystalline Powder
  • pKa: 2.0, 2.7, 6.2, 10.3 (Lit.)
  • Soluble in water : 0.1 M, clear, colorless aqueous solution.

Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Schmelzpunkt: 238…248 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150037
CAS-Nummer: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
pH (1% aq soln) 4-6
Loss on Drying ≤10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR