Guanidiniumchlorid ≥99%, weißes kristallines Pulver für die Molekularbiologie

Lieferant: MP Biomedicals
Achtung

Synonyme: Carbamidin Hydrochlorid, Guanidin-Chlorid, Guanidinhydrochlorid, Aminomethanamidin Hydrochlorid, Aminomethansäureamidiniumchlorid

0219482625 0219482680 0219482690 0219482691 0219482692
ICNA0219482625EA 43.1 EUR
ICNA0219482625 ICNA0219482680 ICNA0219482690 ICNA0219482691 ICNA0219482692
Guanidiniumchlorid ≥99%, weißes kristallines Pulver für die Molekularbiologie
Guanidiniumchlorid

Storage: Room Temperature
Guanidine hydrochloride consists of a network of guanidinium cations and chloride anions linked by N–H•••Cl hydrogen bonds, it is a strong chaotropic agent. Guanidine HCl may agglomerate upon storage. It may appear as a free-flowing crystalline powder, a free flowing powder with solid material dispersed throughout, or a solid. The quality of the product does not appear to be affected and solutions prepared from the free-flowing and lumpy guanidine HCl appear identical.
Guanidine hydrochloride can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary. Also used in the isolation of RNA. Guanidine hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of protein, it can solubilize insoluble or denatured proteins such as inclusion bodies and be used for the recovery of periplasmic proteins.


  • Molecular Biology Reagent Grade
  • Presentation: White Crystalline Powder

Formel: NH₂C(=NH)NH₂·HCl
Schmelzpunkt: 180…190 °C
Dichte: 1,345 g/cm³ (20 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013026
CAS-Nummer: 50-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
Melting Point 180 - 190°C
FTIR Conforms to Standard
pH (6.0 M aq soln) 4-6
OD260 (6.0 M aq soln) ≤0.03
Loss on Drying ≤1.0%
Solubility (6.0 M aq soln) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR