ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat]), hellgrünes Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt
2,2-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonic acid) (ABTS) has been used as a chromogenic substrate for horseradish peroxidase (HRP), both in general activity assays and in ELISA applications. Activity of HRP using ABTS appears about four-fold higher than using pyrogallol. It is mainly used as a substrate in sensitive peroxidase assays.
2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is green in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 1% sodium dodecyl sulfate (SDS). Recommended for ELISA (microwell) procedures, not recommended for membrane applications.
- Presentation: Light Green Powder
- Extinction Coefficient (E1%): EmM(342 nM) > 37.4; EmM(225 nM) > 48.15; EmM(414 nM) = 36 (Lit.)
- Soluble in water (5% w/v - clear, pale green solution).
- Storage temperature: Store at +4 °C. Store Under Nitrogen. Protect from light.
Formel:
C₁₈H₂₄N₆O₆S₄ Molecular Weight: 548,68 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00010404 CAS-Nummer: 30931-67-0 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Standard |
UV/VIS: I max (water) | 340 - 342, 224 - 228 nm |
Moisture | ≤5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...