Bromkresolpurpur, Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
5′,5''-Dibrom-o-kresolsulfonphthalein
Bromocresol Purple is used as a pH indicator (5.2-6.8 : yellow to blue-purple). Used in spectrophotometric determination of serum-albumin.
Bromocresol purple binds quantitatively with human serum albumin forming a stable complex with absorbance maximum at 600 nm. The intensity of the color produced is directly proportional to the albumin concentration in the sample.
Soluble in water (20 mg/ml), ethanol (80 mg/ml) or methyl cellosolve (300 mg/ml). Soluble in methanol (4 mg/ml).
Formel:
C₂₁H₁₆Br₂O₅S Schmelzpunkt: 240 °C (Zers., Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011681 CAS-Nummer: 115-40-2 |
Spezifikation Testergebnisse
Identity Test | Passes |
UV/Vis: l max (MeOH) | 418 ± 3 nm |
pH Color Interval | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...