Cobalt(II)chlorid Hexahydrat, rötlich violette Kristalle

Lieferant: MP Biomedicals
Gefahr

0219510725 0219510780 0219510783
ICNA0219510725EA 21.5 EUR
ICNA0219510725 ICNA0219510780 ICNA0219510783
Cobalt(II)chlorid Hexahydrat, rötlich violette Kristalle
Cobalt(II)chlorid Hexahydrat

Cobalt chloride is utilized in commercial applications such as electroplating, catalyst preparation, painting on glass and porcelain, and in the manufacture of vitamin B12.


  • Key Applications : Indicator for water in dessicants
  • Application Areas : Chemistry
  • Product Type : Biochemicals
  • Biochemical Category : Labels, Dyes & Stains
  • Chemical Class : Labels, Dyes & Stains
  • Vapor Pressure : 40 mm Hg(Lit.)
  • Presentation : Redish Purple Crystals
  • Format : Crystals
  • Solubility : Soluble in water (100 mg/mL), alcohols, acetone, ether, glycerol.


It is also used as a humidity and water indicator; when cobalt chloride is used with desiccants, it will change from blue to pink upon absorption of moisture. Cobalt chloride is also used as an additive in fertilizer and feed.


Solubility : Soluble in water (100 mg/ml), alcohols, acetone, ether, glycerol.

Formel: CoCl₂·6H₂O
Dichte: 1,920 g/mL (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149652
CAS-Nummer: 7791-13-1
UN: 3077
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
pH (0.2 M aq soln) Report Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR