EDTA Dinatriumsalz Dihydrat ≥99%, weißes Pulver

Lieferant: MP Biomedicals

0219517380 0219517383 0219517390 0219517391
ICNA0219517380EA 33.5 EUR
ICNA0219517380 ICNA0219517383 ICNA0219517390 ICNA0219517391
EDTA Dinatriumsalz Dihydrat ≥99%, weißes Pulver
EDTA Dinatriumsalz Dihydrat

Ethylenediamine tetraacetic acid is a polyamino carboxylic acid hexadentate ligand and a chelating agent.


Ethylenediamine tetraacetic acid is a chelator of divalent cations. It also inhibits enzymes, such as metalloproteases, that require divalent cations for activity

Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL: MFCD00150037
CAS-Nummer: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
Melting Point 238 - 248 °C
FTIR Conforms to Standard
pH (1% aq soln) 4 - 6
Moisture ≤10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR