Kaliumdihydrogenphosphat, wasserfrei ≥99,0%, kristallines Pulver

Lieferant: MP Biomedicals

02195453.1 02195453.5
ICNA02195453.1EA 23 EUR
ICNA02195453.1 ICNA02195453.5
Kaliumdihydrogenphosphat, wasserfrei ≥99,0%, kristallines Pulver
Kaliumdihydrogenphosphat

Potassium phosphate is used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.


Soluble in water; insoluble to slightly soluble in ethanol; insoluble in alcohol.


Bitte beachten Research Use Only (RUO).

Formel: KH₂PO₄
MDL: MFCD00011401
CAS-Nummer: 7778-77-0

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Assay ≥99.0%
pH (5% aq soln) 4.1 - 4.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR