Rifampicin, orangenes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: 5,6,9,17,19,21-Hexahydroxy-23-methoxy- 2,4,12,16,18,20,22-heptamethyl-8-[N-(4-methyl- 1-piperazinyl)formimidoyl]-2,7-(epoxypentadeca- [1,11,13]trienimino)-naphtho[2,1-b]furan- 1,11(2H)-dion-21-acetat, Rifampin

0219549005 02195490.1 02195490.2 0219549001 0219549025
ICNA0219549005EA 378 EUR
ICNA0219549005 ICNA02195490.1 ICNA02195490.2 ICNA0219549001 ICNA0219549025
Rifampicin, orangenes Pulver
Rifampicin

Rifampin is a bactericidal antibiotic drug of the rifamycin group. It specifically inhibits DNA-dependent bacterial RNA Polymerase. Mammalian RNA polymerase is not affected.


It is used to treat Tuberculosis and Tuberculosis-related mycobacterial infections. It is widely used as an antipruritic agent in the autoimmune cholestatic liver disease, primary biliary cirrhosis (PBC). It has been shown to cause hepatitis.


Mode of Action: Inhibits initiation of RNA synthesis by binding to β-subunit of RNA polymerase. It Inhibits the assembly of DNA and protein into mature virus particles. EmM (max absorbance, phosphate buffer,pH 7,38): 33,20 (237 nm); 32,10 (255 nm); 27,00 (334 nm);15,40 (475 nm)(Lit.). pKa (in water):1,7 (4-hydroxyl group), 7,9 (4-piperazine nitrogen); in methylcellosolve-water (4:1): 3,6 (4-hydroxyl group), 6,7 (3-piperazine nitrogen)(Lit.).


Soluble in chloroform(349 mg/ml, 25 °C), DMSO(~100 mg/ml), ethyl acetate(108 mg/ml, 25 °C), methanol(16 mg/ml, 25 °C) tetrahydrofuran.

Formel: C₄₃H₅₈N₄O₁₂
Schmelzpunkt: 183…188 °C (Zers., Lit.)
MDL: MFCD00151389
CAS-Nummer: 13292-46-1
EINECS: 236-312-0
Merck Index: 12,08382

Bestellen

Spezifikation Testergebnisse

Synonyms Rifamycin AMP; Rifampicin; 3-(4-Methylpiperazinyliminomethyl)rifamycin SV; Rifampicin
Form Orange Powder
Storage Store at +4 °C
Isoelectric Point 4,8 (in water) (Lit.)
Loss on Drying ≤2%
Optical Rotation [α]D25°=+10,6° (c=0,5% in CDCl3)(Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR