Tetrabutylammoniumdihydrogenphosphat ∼ 97%, Pulver

Lieferant: MP Biomedicals

Synonyme: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

0219551301 0219551305
ICNA0219551301EA 63.5 EUR
ICNA0219551301 ICNA0219551305
Tetrabutylammoniumdihydrogenphosphat ∼ 97%, Pulver
Tetrabutylammoniumdihydrogenphosphat

Ion-associating reagent for HPLC


  • Ion-associating reagent for HPLC


Soluble in water (clear to hazy).


Bitte beachten Research Use Only (RUO).

Formel: C₁₆H₃₈NO₄P
Lagertemperatur: Kühlschrank
MDL: MFCD00064526
CAS-Nummer: 5574-97-0

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ~97%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR