Tetrabutylammoniumdihydrogenphosphat ∼ 97%, Pulver
Lieferant: MP Biomedicals
Synonyme:
Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate
0219551301
0219551305
ICNA0219551301EA
63.5
EUR
ICNA0219551301
ICNA0219551305
Tetrabutylammoniumdihydrogenphosphat ∼ 97%, Pulver
Tetrabutylammoniumdihydrogenphosphat
Ion-associating reagent for HPLC
- Ion-associating reagent for HPLC
Soluble in water (clear to hazy).
Bitte beachten Research Use Only (RUO).
Formel:
C₁₆H₃₈NO₄P Lagertemperatur: Kühlschrank |
MDL:
MFCD00064526 CAS-Nummer: 5574-97-0 |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ~97% |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...