Vancomycin Hydrochlorid (aus Streptomyces orientalis)

Lieferant: MP Biomedicals
Achtung

Synonyme: (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement

02195540.1 02195540.2 0219554001 0219554005
ICNA02195540.1EA 69.6 EUR
ICNA02195540.1 ICNA02195540.2 ICNA0219554001 ICNA0219554005
Vancomycin Hydrochlorid (aus Streptomyces orientalis)
Vancomycin Hydrochlorid

Vancomycin hydrochloride inhibits bacterial mucopeptide biosynthesis.


  • Potency approx. 1000 µg/mg
  • pH 2,5 to 4,5 (5% aqueous solution)
  • Storage temperature +4 °C
  • pH: 2.5 - 4.5 (5% aq solution)

Formel: C₆₆H₇₅Cl₂N₉O₂₄·HCl
Lagertemperatur: Kühlschrank
MDL: MFCD03613611
CAS-Nummer: 123409-00-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR