Vancomycin Hydrochlorid (aus Streptomyces orientalis)

Lieferant: MP Biomedicals
Achtung

02195540.1 02195540.2 0219554001 0219554005
ICNA02195540.1EA 69.6 EUR
ICNA02195540.1 ICNA02195540.2 ICNA0219554001 ICNA0219554005
Vancomycin Hydrochlorid (aus Streptomyces orientalis)
Vancomycin Hydrochlorid
Formel: C₆₆H₇₅Cl₂N₉O₂₄·HCl
Lagertemperatur: Kühlschrank
MDL: MFCD03613611
CAS-Nummer: 123409-00-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR