Eosin G-Lösung 10%

Lieferant: MP Biomedicals

02195700.5 0219570001 0219570010 02195700.1
ICNA02195700.5EA 69.7 EUR
ICNA02195700.5 ICNA0219570001 ICNA0219570010 ICNA02195700.1
Eosin G-Lösung 10%
Eosin G

Eosin Y disodium salt is used in combination with Azure II to show morphological details of marrow cells; buffered Azure A/Eosin Y method for demostrating cell granules, microorganisms and nuclei; for showing insect musculature; for demonstrating cells infected with mosquito-iridescent virus; use in combination with Alum Hematoxylin/Azure II to show mast cells, early necrobiotic changes in cells and protozoans, rickettsias and bacteria in tissues; to demonstrate volutin granules in any microorganism; Bond's modification of Mann's Eosin-Methyl Blue for Negri bodies in smears; Harris stain for Negri bodies in smears; Mann's method for staining Negri bodies in sections; Gridley's stain for Endamoeba in tissue sections; a modified methenamine-silver nitrate method for the demonstration of Endamoeba in sections. It is used as a counterstain for hematoxylin.


Eosin Y, a red xanthene dye, has been noted to reduce the affinity of Mg2+- ATPase for ATP and increase the enzyme affinity for Mg2+ at concentrations of 10-50 μM. At higher concentrations near 100 μM the affinity for Mg2+- ATPase for the ion-activator has been shown to be reduced. This compound has also been reported to be a photosensitizer that works as a molecular photoelectrode via catalyzation of the visible-light-driven electron-transfer reaction. This agent also enhances fluorescent quenching abilities by forming a complex with polyvinulpyrrolidone.

Formel: C₂₀H₆Br₄Na₂O₅
Dichte: 1,015 g/mL (25 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005040
CAS-Nummer: 17372-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR