Creatinphosphat Dinatriumsalz, Hyrdrat ≥98%, Pulver

Lieferant: MP Biomedicals

Synonyme: Sodium creatine phosphate dibasic, N-Methyl-N-(N-phosphonocarbamimidoyl)glycine, disodium salt, Sodium creatine phosphate, Phosphocreatine disodium salt

0219923505
ICNA0219923505EA 89 EUR
ICNA0219923505
Creatinphosphat Dinatriumsalz, Hyrdrat ≥98%, Pulver
Creatinphosphat Dinatriumsalz

Creatine phosphate is substrate for CPK determination.


  • Presentation: White Powder


Soluble in water (150 mg/ml-clear, colorless solution)

Formel: C₄H₈N₃O₅P·2Na
Molecular Weight: 255,077 g/mol
MDL: MFCD00014351
CAS-Nummer: 922-32-7
EINECS: 213-074-6

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
pH (1% aq soln) 7 - 9
Moisture 22±3%
Transmittance 99.10%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR