AlbumiNZ™ Bovine serum albumin (BSA), MP Biomedicals
Lieferant: MP Biomedicals
AlbumiNZ™ Protease-reduced BSA is useful for immunoassays where proteases may degrade proteins or where peroxidase or alkaline phospahatase may hydrolyse substrates.
AlbumiNZ™ Protease-reduced BSA is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality. There is no detectable enzymatic activity.
Biochem/physiol actions: Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.