Cod liver oil, klargelbe Flüsigkeit

Lieferant: MP Biomedicals

0290140501 0290140504
ICNA0290140501EA 65.6 EUR
ICNA0290140501 ICNA0290140504
Cod liver oil, klargelbe Flüsigkeit
Cod liver oil

Cod liver oil is a nutritional supplement derived from liver of cod fish, it has high levels of the omega-3 fatty acids, EPA and DHA.


  • Vitamin A = >850 USP u/gm
  • Vitamin D = >85 USP u/gm


Cod liver oil is a source of omega-3 fatty acids.


Slightly soluble in alcohol; freely soluble in chloroform, ether, carbon disulphide, ethyl acetate and petrolium ether.

Lagertemperatur: Raumtemperatur
MDL: MFCD00146493
CAS-Nummer: 8001-69-2

Bestellen

Spezifikation Testergebnisse

Identity test Passes
Vitamin A ≥850 USP u/gm
Vitamin D ≥85 USP u/gm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR