Baumwollsamenöl, Flüssigkeit
Lieferant: MP Biomedicals
0290141901
ICNA0290141901EA
77.3
EUR
ICNA0290141901
Baumwollsamenöl, Flüssigkeit
Cottonseed oil
Soluble in 400 mg plus 4 ml of chloroform - clear, faint yellow to yellow-green.
- Iodine Value: 109 - 120
- Saponification Number: 190-198 (Lit.)
Soluble in 400 mg plus 4 ml of chloroform - clear, faint yellow to yellow-green.
Dichte:
0,92 g/ml (25 °C) (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00130872 CAS-Nummer: 8001-29-4 |
Spezifikation Testergebnisse
Identity Test | Passes |
Source | Gossypium hirsutum |
Myristic (C-14:0) | 0,5 - 2,0% |
Palmitic (C-16:0)% | 17 - 29 |
Steric (C-18:0) | 1,0 - 4,0% |
Oleic (C-18:1) | 13 - 44% |
Linoleic (C-18:2) | 40 - 63% |
Linolenic (C-18:3) | 0,1 - 2,1% |
Iodine Value | 109 - 120 |
Specific Gravity | 0,915 - 0,921 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...