D(+)-Glucose, wasserfrei, weißes Pulver

Lieferant: MP Biomedicals

0290152101 0290152105 0290152110 0290152125
ICNA0290152101EA 138 EUR
ICNA0290152101 ICNA0290152105 ICNA0290152110 ICNA0290152125
D(+)-Glucose, wasserfrei, weißes Pulver
D(+)-Glucose

Dextrose is a simple sugar that is the primary source of energy for the body.


D-(+)-Dextrose anhydrous is used in study of dietary composition specifies consumption, obesity and lifespan in Drosophila melanogaster.

Formel: C₆H₁₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00148912
CAS-Nummer: 50-99-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
pH (10% aq soln) 4,6 - 6,5
Melting Point 150 - 170° C
Loss on Drying ≤0,5% max
Optical Rotation (c=10, H₂O + 1 drop NH₄OH) Report result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR