Brewers yeast, MP Biomedicals
Lieferant: MP Biomedicals
0290331205
0290331225
0290331280
ICNA0290331205EA
84.7
EUR
ICNA0290331205
ICNA0290331225
ICNA0290331280
Brewers yeast, MP Biomedicals
Zellkultur-Ergänzungen und -Zusätze
Brewers yeast is often used as a source of B-complex vitamins, chromium, and selenium. It is also can be used to bake bread, ferment wine, beer and ethanol production.