Fish oil, from menhaden, klargelbe Flüsigkeit, raffiniert

Lieferant: MP Biomedicals

0296012001
ICNA0296012001EA 129 EUR
ICNA0296012001
Fish oil, from menhaden, klargelbe Flüsigkeit, raffiniert
Fish oil, from menhaden

Menhaden fish oil has been refined to produce a more palatable, clearer oil. It has undergone the winterizing process similar to the alkali refined and bleached oil. The cold test results in a minimum of two hours at 40 °F.


Menhaden Oil is rich in omega-3 fatty acids. When used to partially supplement a high-fat diet in diabetic rats. Menhaden oil improved end points for peripheral neuropathy by improving motor and sensory nerve conduction velocity. Decreased accumulation of fatty acids in rat livers was also demonstrated.


Menhaden oil is use as a control or check sample for fatty acid methyl ester (FAME) analyses.


Bitte beachten Store at -20 °C, store under nitrogen.

Lagertemperatur: Gefrierschrank
MDL: MFCD00131085
CAS-Nummer: 8002-50-4

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Typical Fatty Acid Composition (% wt.) for C20:5 14 ± 3%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR