Bisacrylamid 99, powder_white, ultrapure

Lieferant: MP Biomedicals
Achtung

Synonyme: N,N'-Methylendiacrylamid, BIS

04800172
ICNA04800172EA 44.8 EUR
ICNA04800172
Bisacrylamid 99, powder_white, ultrapure
Bisacrylamid

N,N'-Methylene-bis-acrylamide is one of the compounds of the polyacrylamide gel. Bisacrylamide polymerises with acrylamide and is capable of creating cross-links between polyacrylamide chains, thus creating a network of polyacrylamide.


  • Acrylic Acid Content ≤0.02%


N,N'-Methylene-bis-acrylamide is one of the compounds of the polyacrylamide gel. Bisacrylamide polymerises with acrylamide and is capable of creating cross-links between polyacrylamide chains, thus creating a network of polyacrylamide.


N,N'-Methylene-bis-acrylamide is a cross-linker reagent used for precise, critical PAGE gels. May be used in UV scanning gels. It should mixed with acrylamide for making polyacrylamide gels for use in protein and nucleic acid electrophoresis.


Soluble in 2% water (20 mg/ml at 20 °C).

Formel: (H₂C=CHCONH)₂CH₂
Molecular Weight: 154.17 g/mol
Siedepunkt: 125
Schmelzpunkt: 185
Dichte: 1.235 g/cm³ (20 °C)
Flammpunkt: 138
Lagertemperatur: Raumtemperatur
MDL: MFCD00008625
CAS-Nummer: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Synonyms bis-Acrylamide; Methylenebisacrylamide; N,N'-Methylenediacrylamide; BIS
Form White Powder
Storage Store at room temperature (15 to 30 °C), protect from light, store under nitrogen
Conductivity ≤10 µmhos
pH ≥5 (2,5% aq soln)
Acrylic acid content ≤0,02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR