Methylenblau Trihydrat, Pulver C.I. 52015
Lieferant: MP Biomedicals
Synonyme:
3,7-Bis(dimethylamino)phenothiaziniumchlorid Trihydrat, N,N,N′,N′-Tetramethylthioninchlorid Trihydrat, Methylthioniniumchlorid Trihydrat, C.I.52015
Methylene blue has been used as a stain in bacteriology and as an oxidation-reduction indicator. A sensitive stain for RNA and
ribonuclease. An ethidium bromide alternative for the visualization of nucleic acids in gels:
- Soluble in water (1 mg/ml)
Gels are viewable in visible light. Does not require a darkroom. No UV induced DNA damage. Gels can be stored for several days. 30 minute destaining time for PAGE gels.
Soluble in water (1 mg/ml)
Bitte beachten Research Use Only (RUO).
Formel:
C₁₆H₁₈ClN₃S·3H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150008 CAS-Nummer: 7220-79-3 |
Spezifikation Testergebnisse
Identity Test | Passes |
UV/Vis: λmax (water) | 662 ± 3 nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...