Methylgrün-Zinkchlorid-Doppelsalz, dunkles Pulver
Lieferant: MP Biomedicals
Methyl green may be used as a pH indicator. Methyl green solution is a suitable counterstain for chloroacetate esterase, nonspecific esterase, alkaline phosphatase, peroxidase, naphthol AS acetate esterase, and acid phosphatase
- Soluble in water
Formel:
C₂₇H₃₅BrClN₃·xZnCl₂ |
MDL:
MFCD00151094 CAS-Nummer: 7114-03-6 |
Spezifikation Testergebnisse
Synonyms | CI 42590; 4-[[4-(Dimethylamino) phenyl][4-(dimethyliminio) cyclohexa-2,5-dien-1-ylidene] methyl]- N-ethyl-N,N- bromide chloride, compound with zinc chloride; Ethyl Green |
Form | Dark Powder |
Storage | Store at Room Temperature (15 - 30 °C) |
Color Index | 42590 |
Extinction Coefficient | E1% = 800 - 1100 (water) Lit. |
Typical Working Concentration | A typical working concentration for using methyl green as a counter stain is 0,1 - 0,5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...