Methylgrün-Zinkchlorid-Doppelsalz, dunkles Pulver

Lieferant: MP Biomedicals

04806404 04806408
ICNA04806404EA 53.3 EUR
ICNA04806404 ICNA04806408
Methylgrün-Zinkchlorid-Doppelsalz, dunkles Pulver
Methylgrün-Zinkchlorid-Doppelsalz

Methyl green may be used as a pH indicator. Methyl green solution is a suitable counterstain for chloroacetate esterase, nonspecific esterase, alkaline phosphatase, peroxidase, naphthol AS acetate esterase, and acid phosphatase


  • Soluble in water

Formel: C₂₇H₃₅BrClN₃·xZnCl₂
MDL: MFCD00151094
CAS-Nummer: 7114-03-6

Bestellen

Spezifikation Testergebnisse

Synonyms CI 42590; 4-[[4-(Dimethylamino) phenyl][4-(dimethyliminio) cyclohexa-2,5-dien-1-ylidene] methyl]- N-ethyl-N,N- bromide chloride, compound with zinc chloride; Ethyl Green
Form Dark Powder
Storage Store at Room Temperature (15 - 30 °C)
Color Index 42590
Extinction Coefficient E1% = 800 - 1100 (water) Lit.
Typical Working Concentration A typical working concentration for using methyl green as a counter stain is 0,1 - 0,5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR