Polysorbat 20, klargelbe Flüsigkeit

Lieferant: MP Biomedicals

Synonyme: Sorbitanmonolaurat, ethoxyliert, Sorbitanmonododecanoat, ethoxyliert, Polyoxyethylen-sorbitan-monolaurat, Polyethylenglycol-sorbitan-monolaurat

04806576
ICNA04806576EA 30.8 EUR
ICNA04806576
Polysorbat 20, klargelbe Flüsigkeit
Polysorbat 20

Nonionic surfactant derived from sorbitan esters, widely used in biochemical applications.


  • Saponification Number 40 - 50
  • complies for IR spectroscopy


Polysorbate 20 is used as emulsifying agent for the preparation of stable oil-inwater emulsions in pharmaceuticals, cosmetics, cleaning compounds. Polysorbate 20 has been used in preextraction of membranes to remove peripheral proteins (used at 2% for extraction of membrane-bound proteins). It has been used as a blocking agent for membrane based immunoassays at a typical concentration of 0.05%. It can be used for lysing mammalian cells at a concentration of 0.05 to 0.5%.


Soluble/miscible in water to give a clear yellow solution; miscible with alcohol, dioxane, and ethyl acetate; practically
insoluble in liquid paraffin and fixed oils (such as mineral oil)


Bitte beachten Research Use Only (RUO).

Formel: C₅₈H₁₁₄O₂₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00165986
CAS-Nummer: 9005-64-5

Bestellen

Spezifikation Testergebnisse

Identity Test A Passes
Identity Test B Passes
FTIR Conforms to Reference
Solubility (1 g/20 ml) Clear, Foamy Solution
Specific Gravity 1.08 - 1.13
Water Content ≤3%
Saponification Number 40 - 50

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR