Harnstoff ≥99%, weiße Granalien, ultrarein

Lieferant: MP Biomedicals

Synonyme: Urea, Aminoameisensäureamid, Carbamid, Carbonyldiamid

04821519 04821527 04821528 04821530 04821532
ICNA04821519EA 55.3 EUR
ICNA04821519 ICNA04821527 ICNA04821528 ICNA04821530 ICNA04821532
Harnstoff ≥99%, weiße Granalien, ultrarein
Harnstoff

Urea is the principal end product of nitrogen metabolism in most mammals, formed by the enzymatic reactions of the Kreb's cycle.


  • Ultra Pure Grade
  • Partition Coefficient: log Pow: -2,59 - -1,59
  • One gram dissolves in 1 mL water, 10 mL 95% ethanol, 1 mL boiling 95% ethanol, 20 mL absolute ethanol, 6 mL methanol, 2 mL glycerol; Soluble in concentrated hydrochloric acid; almost insoluble in chloroform, ether.
  • Storage temperature: Room Temperature


Urea is a mild agent usually used in the solubilization and denaturation of proteins. It is also useful for renaturing proteins from samples already denatured with 6 M guanidine hydrochloride such as inclusion bodies; and in the extraction of the mitochondrial complex. It is commonly used to solubilize and denature proteins for denaturing isoelectric focusing and two-dimensional electrophoresis and in acetic acid-urea PAGE gels.


Urea is used in cell or tissue culture media to increase the osmolality. Urea has also been used as fertilizer because of the easy availability of nitrogen; in animal feeds; it is reacted with aldehydes to make resins and plastics; condensed with malonic ester to form barbituric acid; used in the paper industry to soften cellulose; used as a diuretic; enhances the action of sulfonamides; an antiseptic.


Urea in solution is in equilibrium with ammonium cyanate. The form that reacts with protein amino groups is isocyanic acid. Urea in the presence of heat and protein leads to carbamylation of the proteins. Carbamylation by isocyanic acid interferes with protein characterization because isocyanic acid reacts with the amino terminus of proteins, preventing N-terminal sequencing. Isocyanic acid also reacts with side chains of lysine and arginine residues resulting in a protein that is unsuitable for many enzymatic digests. In addition, carbamylation often leads to confusing results from peptides having unexpected retention times and masses. When performing enzymatic protein digests it is important to remove urea first. Even though some enzymes will tolerate small amounts of urea, the elevated temperature used for most reactions will lead to carbamylation during the course of the digest. The urea can be removed prior to digestion by fast reversed phase chromatography, spin columns, or dialysis.


Dissolve urea in deionized water to the desired concentration.For every 10 ml of solution, add 1 g of Amberlite® IRA-910.Stir for one hour at room temperature

Formel: NH₂CONH₂
Siedepunkt: 135 °C (Zersetzung)
Dichte: 1,335 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008022
CAS-Nummer: 57-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR