Coomassie® Brillantblau R 250, dunkles Pulver für die Elektrophorese

Lieferant: MP Biomedicals

Synonyme: Brillantblau R, Hydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylen]cyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium Mononatriumsalz, Coomassie Blau R 250, Xylenbrillantcyanin, Serva Blue R, Brillantindocyanin 6B, Acid Blue 83

04821616 04821636
ICNA04821616EA 69.9 EUR
ICNA04821616 ICNA04821636
Coomassie® Brillantblau R 250, dunkles Pulver für die Elektrophorese
Coomassie® Brillantblau R 250

Coomassie® Brilliant Blue R-250 is a fast acting, sensitive dye, which can be used on SDS gels, IEF gels, and standard gels.


  • For detection of protein bands following electrophoresis


Brilliant Blue R can be used for staining proteins on polyacrylamide electrophoresis gels. Improvement of sensitivity and background have been accomplished using Brilliant Blue dyes in a colloidal form. Other applications for brilliant Blue R include use in detection of lipids in thin layer chromatography, determination of the CMC of detergents, and detection of proteins in solution, as well as on cellulose acetate plates.


Soluble in water (10 mg/ml), 20 mg/ml in methyl cellosolve, ethanol (10 mg/ml), methanol.


Bitte beachten Research Use Only (RUO).

Formel: C₄₅H₄₄N₃NaO₇S₂
Molecular Weight: 825.98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00041762
CAS-Nummer: 6104-59-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
λmax (ethanol) 585 ± 5 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR