Decontamination solution, DNA-Erase™, MP Biomedicals
Lieferant: MP Biomedicals
DNA-Erase™ is a non-alkaline, non-corrosive and non-carcinogenic cleansing solution that is highly active against DNA contamination and is intended for use at PCR workstations for eliminating PCR artifacts from all laboratory surfaces, including benchtops, apparatus and pipettors.
- Soluble in water
- Stable and heat resistant
- Ready to use
DNA-Erase™ solution contains a surfactant plus an agent that destroys DNA.