Alizarin-Komplexon

Lieferant: MP Biomedicals

0520342901
ICNA0520342901EA 106 EUR
ICNA0520342901
Alizarin-Komplexon
Alizarin-Komplexon
Formel: C₁₉H₁₅NO₈
Schmelzpunkt: 180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001202
CAS-Nummer: 3952-78-1
EINECS: 223-544-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR