Ammonium succinate, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: Butanedioic acid, diammonium salt, Diammonium succinate, Succinic Acid Diammonium Salt

0520504980
ICNA0520504980EA 905 EUR
ICNA0520504980
Ammonium succinate, weißes Pulver
Ammonium succinate
Formel: C₄H₁₂N₂O₄
Molecular Weight: 152,15 g/mol
Schmelzpunkt: 165 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00020534
CAS-Nummer: 2226-88-2

Bestellen

Spezifikation Testergebnisse

Synonyms Succinic acid diammonium salt
Molecular formula C4H12N2O4
Molecular weight 152,15 g/mol
CAS number 2226-88-2
Form Crystalline powder
Colour White
Flammability (solid, gaseous) Non-flammable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR