Magnesiumchlorid, anhydrous, powder_white crystalline

Lieferant: MP Biomedicals

0520984480
ICNA0520984480EA 44.3 EUR
ICNA0520984480
Magnesiumchlorid, anhydrous, powder_white crystalline
Magnesium chloride

Magnesium chloride is a widely used reagent in chemistry and molecular biology as a source of magnesium ion.


Synonyms: Magnogene, chloromagnestite.

Formel: MgCl₂
Molecular Weight: 95.21 g/mol
Siedepunkt: 1412
Schmelzpunkt: 715
Lagertemperatur: Raumtemperatur
MDL: MFCD00011106
CAS-Nummer: 7786-30-3
Merck Index: 13,05684

Bestellen

Spezifikation Testergebnisse

Appearence White anhydrous crystalline powder
Solubility Soluble in exothermic, water (4 g/10 ml-clear colourless haze free solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR