Lithiumcobaltit (Lithiumcobalt(III)oxid)

Lieferant: MP Biomedicals

Synonyme: Lithium cobalt(III) oxide

0522108580
ICNA0522108580EA 91.2 EUR
ICNA0522108580
Lithiumcobaltit (Lithiumcobalt(III)oxid)
Lithiumcobaltit (Lithiumcobalt(III)oxid)
Formel: CoLiO₂
Molecular Weight: 97,87 g/mol
MDL: MFCD00049786
CAS-Nummer: 12190-79-3
EINECS: 235-362-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR