Streptomyces avidini Streptavidin, MP Biomedicals
Lieferant: MP Biomedicals
08623001
ICNA08623001EA 65.3 EUR
ICNA08623001
Streptomyces avidini Streptavidin, MP Biomedicals
Proteine und Peptide
Streptavidin effectively binds to biotin with an isoelectric point close to neutral and it contains no carbohydrate residues.
Unit definition: One unit of streptavidin will bind 1 µg of biotin.