Freund's incomplete adjuvant, MP Biomedicals
Lieferant: MP Biomedicals
08642861
ICNA08642861EA
119
EUR
ICNA08642861
Freund's incomplete adjuvant, MP Biomedicals
Protein- Expression
Proteinexpressions-Reagenzien
Freund's incomplete adjuvant mixture contains mineral oil and Arlacel A.
- Enhances immune response in booster injections
- Water-in-oil emulsion without Mycobacterium